missing translation for 'onlineSavingsMsg'
Få mere at vide

TOKU-E™ Spectinomycin ReadyMade™ Solution

Artikelnummer. 30044056
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 x 1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30044056 10 x 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30044056 Leverandør TOKU-E™ Leverandørnr. S13710x1ML
 Begrænset vare
Forventet forsendelse: 07-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Spectinomycin ReadyMade™ Solution is provided as a sterile-filtered solution of Spectinomycin Sulfate in water.

  • CAS Number: 64058-48-6
  • Molecular Formula: C14H26N2O11S · H2SO4 · 4H2O
  • Molecular Weight: 502.49 g/mol
  • Storage Conditions: -20°C, Protect from light
  • Form: Liquid
  • Source: Streptomyces sp.
  • Spectrum: Spectinomycin Sulfate is broad-spectrum antibiotic active against a variety of Gram-negative and Gram-positive organisms. Anaerobic organisms are generally resistant. Spectinomycin is usually bacteriostatic
  • Mechanism of Action: The aminocyclitol antibiotic Spectinomycin, often considered alongside the aminoglycosides, binds in reversible fashion (hence the bacteriostatic activity) to the 16S rRNA of the ribosomal 30S subunit. There it interrupts the translocation event that occurs as the next codon of mRNA is aligned with the A site in readiness for the incoming aminoacyl-tRNA. Structural studies reveal that the antibiotic binds to an area of the 30S subunit known as the head region which needs to move during translocation. Binding of the rigid Spectinomycin molecule appears to prevent the movement required for translocation

Tekniske data

CAS 64058-48-6
Certificeringer/compliance Assay: Spectinomycin Sulfate on dried basis: 93 to 102%
Kemisk navn eller materiale Spectinomycin Sodium
Farve Clear, colorless
Indhold og opbevaring -20°C
Oversigt Spectinomycin ReadyMade™ Solution 10mg/mL in Water
Koncentration 100 mg/mL
Form Clear, colorless solution
Produkttype Antibiotic
Sterilitet Sterile
Steriliseringsmetode Sterile-filtered
Med tilsætningsstoffer None
Uden tilsætningsstoffer None
Formulering Liquid in Water
Til brug med (applikation) Microbiology, Plant Biology, Eukaryotic Cell Culture and Cancer Applications
Produktlinje ReadyMade™ Solutions
Mængde 10 x 1 ml
Forsendelsestilstand Ice Pack
Kilde Streptomyces sp.
Arter Bacteria
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.