missing translation for 'onlineSavingsMsg'
Learn More
Spebrutinib, MedChemExpress Product Code.: 16477416

Spebrutinib, MedChemExpress

Product Code. p-200100326
100 mg, 100mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16477416

Brand: Medchem Express HY18012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Spebrutinib (AVL-292; CC-292) is a covalent, orally active, and highly selective with an IC50 of 0.5 nM.

Spebrutinib (AVL-292

Specifications

Chemical Name or Material Spebrutinib
CAS 1202757-89-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H22FN5O3
Quantity 100 mg
Synonym AVL-292 CC-292
Solubility Information DMSO : ≥ 45 mg/mL (106.27 mM)
SMILES COCCOC1=CC=C(NC2=NC=C(F)C(NC3=CC=CC(NC(C=C)=O)=C3)=N2)C=C1
Molecular Weight (g/mol) 423.44
Formula Weight 423.44
Percent Purity 95.15%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Spebrutinib, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.