Få mere at vide
Brady™ SPC Sorbent Center (cabinet only plus 2 racks)
Artikelnummer. 10742997 Shop alle Brady produkter
Skift visning
Dimensions (L x W x H): 564 x 1270 x 660 mm
Artikelnummer.
10742997 Dimensions (L x W x H): 564 x 1270 x 660 mm
1 options
Artikelnummer. 10742997 Leverandør Brady™ Leverandørnr. 813899
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
The first modular and mobile storage cabinet designed specifically for sorbents
  • Made from strong, durable and chemically resistent polyethylene for longer life
  • Double use: use it as a spill kit or as a sorbent storage and dispensing station
  • Modular design: stack 2 or 3 units together in order to ake a cabinet
  • Dual access doors: allows easy and quick access to the product
  • Add wheels and it is mobile
Tekniske data
Type SPC Kits
Emballagemængde 1 stk.
Dimensioner (L x B x H) 564 x 1270 x 660 mm
Højde (metrisk) 660.40 mm
Dybde (metrisk) 546.10 mm
Vægt (metrisk) 22,680 g