Learn More
Brady™ SPC™ Absorbents Universal Refill Kit for Spill Kaddie
Product Code. 30533350 Shop All Brady Products
Change view
Click to view available options
Capacity (English):
15.850 gal.
Unit Size:
Each
Product Code. Capacity (English) Unit Size
30533350 15.850 gal. Each
1 options
Product Code. 30533350 Supplier Brady™ Supplier No. 830753
  • ONLINE EXCLUSIVE PRICE
    2778.90 DKK
    2450.97 DKK / Pakke med 1
    Spare 327.93 DKK
    11% Slukket
 Restricted Item
På lager
Refill kit for SKA-CART.
  • Refill kit for SKA-CART
Specifications
Color Gray
Capacity (English) 15.850 gal.
Type Refill Kit
For Use With (Application) Beverage Processing, Chemical, Food Processing, Laboratory, Manufacturing, Maritime, Oil and Gas, Process, Warehouse and Logistics
Includes 1 Double Perforated Roll (37 cm x 23 m), 2 Socs (7.6 x 122 cm), 1 Soc (7.6 x 366 cm), 6 Pillows (25 x 25 cm), 1 Pair of Gloves, 1 Pair of Goggles and 2 Disposal Bags
Model Universal Sorbent
Alternate Part Number SKA-CART-RF