Få mere at vide
Brady™ SPC™ Absorbents Drain Cover Bags Large
Beskrivelse
- BAG-PU-L is a carry bag made from Polyurethane that protects the PU90, PU110 drain seal during storage and facilitates transport.
Tekniske data
Tekniske data
| Farve | Yellow |
| Type | Drain Cover Bags |
| Til brug med (udstyr) | To Protect the PU90, PU110 Drain Seal or SB-3 Spill Barrier During Storage and to Facilitate Transport |
| Model- | Spill Containment |
| Længde (engelsk) | 4.265 ft. |
| Længde (metrisk) | 1.30 m |
| Diameter (engelsk) | 9.843 in. |
| Diameter (metrisk) | 25.00 cm |
| Alternativt varenummer | BAG-PU-L |