Få mere at vide
(+)-Sparteine, TRC
Artikelnummer. 30132870
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132870 100 mg
30137536 50 mg
30006470 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132870 Leverandør Toronto Research Chemicals Leverandørnr. TRCS679505100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 492-08-0
Molekylær formel C15 H26 N2
Molekylvægt (g/mol) 234.38
Synonym 7,14-Methano-2H,6H-dipyrido[1,2-a:1',2'-e][1,5]diazocine, dodecahydro-, (7R,7aR,14R,14aS)-, 7,14-Methano-2H,6H-dipyrido[1,2-a:1',2'-e][1,5]diazocine, dodecahydro-, [7R-(7α,7aα,14α,14aβ)]-, Pachycarpine (6CI,8CI), (+)-Sparteine, 6α,7β,9β,11β-Sparteine, D-(+)-Sparteine, D-Sparteine, d-Sparteine
SMIL C1CCN2C[C@H]3C[C@H](CN4CCCC[C@@H]34)[C@H]2C1
Tekniske data
Kemisk navn eller materiale (+)-Sparteine
Mængde 100 mg
InChI formel InChI=1S/C15H26N2/c1-3-7-16-11-13-9-12(14(16)5-1)10-17-8-4-2-6-15(13)17/h12-15H,1-11H2/t12-,13-,14-,15+/m1/s1
Formel vægt 234.2096