Få mere at vide
Sparteine Sulfate, TRC
Artikelnummer. 30011274
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30011274 2.5 g 2.5g
30011717 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011274 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6795002.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 299-39-8
Molekylær formel C15 H26 N2 . H2 O4 S
Molekylvægt (g/mol) 332.46
Synonym 7,14-Methano-2H,6H-dipyrido[1,2-a:1',2'-e][1,5]diazocine, dodecahydro-, (7S,7aR,14S,14aS)-, sulfate (1:1), 7,14-Methano-2H,6H-dipyrido[1,2-a:1',2'-e][1,5]diazocine, dodecahydro-, [7S-(7α,7aα,14α,14aβ)]-, sulfate (1:1), Sparteine, sulfate (1:1) (8CI), (-)-Sparteine sulfate, NSC 143087, NSC 26253, NSC 402663, Spal, Spareng, Sparm, Sparteal, Sparteine sulfate, Spartepur, Spartin, Spartocin, Spatym, Tocosimplex
SMIL OS(=O)(=O)O.C1CCN2C[C@@H]3C[C@@H](CN4CCCC[C@@H]34)[C@H]2C1
Tekniske data
Kemisk navn eller materiale Sparteine Sulfate
Mængde 2.5 g
InChI formel InChI=1S/C15H26N2.H2O4S/c1-3-7-16-11-13-9-12(14(16)5-1)10-17-8-4-2-6-15(13)17;1-5(2,3)4/h12-15H,1-11H2;(H2,1,2,3,4)/t12-,13-,14-,15+;/m0./s1
Formel vægt 332.177