Learn More
Merck Accessories for Swinnex Filter Holders
Accessories for filter holders for sample preparation.
Brand: Merck SX00047RK
Additional Details : Weight : 0.10000kg
Description
MilliporeSigma Spare Parts, For Use With: For Swinnex 47, Includes: 10 Grid-together support and 10 boards for the base and 2 Plastic wrenches
Specifications
Not Autoclavable | |
For Swinnex 47 | |
10 Grid-together support and 10 boards for the base and 2 Plastic wrenches |
Spare Parts | |
Non-sterile | |
Laboratory Filter Holders or Cyclones |