Learn More
DWK Life Sciences DURAN™ Replacement 0.2 μm Syringe Disk Filter, for Pressure Equalization Set (GL 45 Connection Cap System Accessory)
Accessory for 2 or 3 port GL 45 connection cap system
Brand: DWK Life Sciences 1129819
114.95 DKK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.01100kg
Description
Flexible modular GL 45 connection cap system that accepts four different rigid or semi-rigid tubing diameters (1.6 mm; 3.0 mm; 3.2 mm and 6.0 mm). The caps are available with 2 or 3 GL 14 threaded ports. The different sized inserts form a tight seal around the pass-through tubing. Sterile pressure equalisation is possible through use of 0.2 micron syringe filter. Unused ports can be sealed off with a red GL 14 screw cap.
- Rigid or semi-rigid tubing of different diameters can be accepted by using a blue GL 14 open topped cap with one of the inserts
- Available with 2, or 3 threaded outlets (ports)
- Five different tubing diameters (3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm and 12.0 mm) can be used by changing the inserts
- By applying positive or negative pressure liquid can be pumped out of or into the bottle via the screw cap
- Fully compatible with all the GL 45 threads and therefore highly versatile
- Fully compatible with all the GLS 80 threads and therefore highly versatile
Suitable Accessory: 1137799
Specifications
Autoclavable | |
Borosilicate Glass (3.3), Polypropylene, Silicone | |
Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer between lab bottles; Supply of reagents to instruments; Bioreactor supply | |
Spare syringe filter for pressure compensation |
GL 14 thread size | |
0.2 μm | |
2/Pk. | |
87 mm |