Learn More
BRAND™ Spare Lateral Stopcock with Key
Stopcock housing with burette tip includes interchangeable PTFE or borosilicate glass key with retention device.
Brand: BRAND™ 81167
266.37 DKK valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
- Lateral stopcock housing with burette tip
- Interchangeable PTFE or borosilicate glass key with retention device
Specifications
50 mL | |
PTFE | |
Retention Device |
Burette | |
1 Each | |
Stopcock Key |