Learn More
RETSCH Spare Collecting Receptacle
Brand:  RETSCH 05.000.0019
508.25 DKK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.20000kg
Product Code. 10719522
  • 535.00 DKK / Each
Estimated Shipment: 10-07-2024
to see stock.
Specifications
Spare Collecting Receptacle
1.5 L
For Sample Splitter RT 6.5 and RT 12.5