Learn More
Eppendorf™ Spare Adaptor Clamp
Brand:  Eppendorf™ 5804781007
70.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10777292
  • 70.28 DKK / Each
Specifications
Adapter Clamp
100mL Bucket
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Eppendorf™ Spare Adaptor Clamp > For 100mL Bucket