Få mere at vide
Spaglumic Acid, TRC
Artikelnummer. 30416857
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416857 50 mg
30422218 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30416857 Leverandør Toronto Research Chemicals Leverandørnr. TRCS67910050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3106-85-2
Molekylær formel C11H16N2O8
Molekylvægt (g/mol) 304.25
Synonym N-Acetyl-L-alpha-aspartyl-L-glutamic Acid, N-(N-Acetyl-L-alpha-aspartyl)-L-glutamic Acid, Acetyl-alpha-L-aspartylglutamic Acid, Isospaglumic Acid, N-(N-Acetylaspartyl)glutamic Acid, N-Acetyl-L-aspartyl-L-glutamic Acid, N-Acetyl-alpha-L-aspartyl-L-glutamic Acid, N-Acetyl-alpha-aspartylglutamic Acid, N-Acetylaspartylglutamic Acid, NAAG, alpha-Spaglumic Acid
IUPAC navn (2S)-2-[[(2S)-2-acetamido-3-carboxypropanoyl]amino]pentanedioic acid
SMIL CC(=O)N[C@@H](CC(=O)O)C(=O)N[C@@H](CCC(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale Spaglumic Acid
Mængde 50 mg
InChI formel InChI=1S/C11H16N2O8/c1-5(14)12-7(4-9(17)18)10(19)13-6(11(20)21)2-3-8(15)16/h6-7H,2-4H2,1H3,(H,12,14)(H,13,19)(H,15,16)(H,17,18)(H,20,21)/t6-,7-/m0/s1
Formel vægt 304.0907