Få mere at vide
Bel-Art™ Sterileware™ Økonomi prøve skeer
Shop alle SP Scienceware produkter
Kapacitet (metrisk):
1.25 mL
2.5 mL
5 mL
8 mL
Pakningsstørrelse:
Pakke med 10
Beskrivelse
- Made from high-impact polystyrene, FDA grade and sterilized by gamma irradiation
- Comes with flat bottom shape to allow use as weighing boat
- Available in white color
- Available in individual packs for one time use
- Includes certificate of processing for sterilization
Tekniske data
Tekniske data
| Sterilitet | Sterile |
| Autoklaverbar | Not Autoclavable |
| Form | Flat Bottom |
| Kapacitet (metrisk) | 2.5 mL |
| Farve | White |
| Certificeringer/compliance | FDA |
| Engangs | Single-use |
| Produkttype | Spoon |
| Til brug med (applikation) | Ideal for retrieving small, specific volume samples of powders or liquids, provides durability and stiffness when sampling packed powders |
| Inkluderer | Certificate of processing for sterilization |
| Vis mere |