missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
SOX4 Antibody (PCRP-SOX4-1D6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14180H
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SOX4 Monoclonal antibody specifically detects SOX4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Tekniske data
| SOX4 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| PCRP-SOX4-1D6 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4 | |
| Recombinant fragment (around aa45-130) of human SOX4 protein (Uniprot: Q06945) | |
| 0.1 mL | |
| Stem Cell Markers, Stem Cells | |
| 6659 | |
| Store at 4°C in the dark. | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion