Learn More
SOX2 Monoclonal Antibody (SP76), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA516399
Additional Details : Weight : 0.01000kg
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is prostate or lung squamous cell carcinoma.
SOX2 Monoclonal antibody specifically detects SOX2 in Human, Mouse samples. It is validated for ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
SOX2 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
P48431, P48432 | |
SOX2 | |
Synthetic peptide corresponding to N-terminus of human SOX-2 protein. | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
SP76 | |
Unconjugated | |
SOX2 | |
ANOP3; cb236; Delta EF2a; lcc; MCOPS3; MCOPS3 (Microphthalmia Syndromic type 3); RGD1565646; sex determining region Y-box 2; SOX 2; SOX2; Sox-2; soxp; SRY (sex determining region Y) box 2; SRY (sex determining region Y)-box 2; SRY box 2; SRY related HMG box 2; SRY-box 2; SRY-box 2; SOX2; SRY-box containing gene 2; SRY-box transcription factor 2; SRY-related HMG-box gene 2; transcription factor SOX2; transcription factor SOX-2; wu:fb83g04; wu:fc14d07; ysb; zgc:65860; zgc:77389 | |
Rabbit | |
Protein A/G | |
RUO | |
20674, 6657 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |