Learn More
Sotalol Hydrochloride, LoGiCal
Product Code. 30457989
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30457989 10 mg 10mg
1 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30457989 Supplier LoGiCal Supplier No. LGCFOR0046.00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sotalol Hydrochloride
CAS 959-24-0
Analyte or Component Names Sotalol Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C12 H20 N2 O3 S . HCl
Certifications/Compliance ISO 9001
Synonym Methanesulfonamide, N-[4-[1-hydroxy-2-[(1-methylethyl)amino]ethyl]phenyl]-, hydrochloride (1:1), Methanesulfonamide, N-[4-[1-hydroxy-2-[(1-methylethyl)amino]ethyl]phenyl]-, monohydrochloride (9CI), Methanesulfonanilide, 4'-[1-hydroxy-2-(isopropylamino)ethyl]-, monohydrochloride (8CI), 4'-[1-Hydroxy-2-(isopropylamino)ethyl]methanesulfonanilide hydrochloride, 4'-[2-(Isopropylamino)-1-hydroxyethyl]methanesulfonanilide hydrochloride, 4'-[2-(Isopropylamino)-1-hydroxyethyl]methanesulfonanilide monohydrochloride, Betapace, DL-MJ 1999, Darob, MJ 1999, N-[4-[1-Hydroxy-2-[(1-methylethyl)amino]ethyl]phenyl]methanesulfonamide hydrochloride, Sotacor, Sotalex, Sotalol HCl, Sotalol hydrochloride, dl-Sotalol hydrochloride, (+/-)-Sotalol hydrochloride, N-Isopropyl-beta-(4-methanesulfonamidophenyl)ethanolamine hydrochloride
InChI Formula InChI=1S/C12H20N2O3S.ClH/c1-9(2)13-8-12(15)10-4-6-11(7-5-10)14-18(3,16)17;/h4-7,9,12-15H,8H2,1-3H3;1H
Solution Type Neat
SMILES Cl.CC(C)NCC(O)c1ccc(NS(=O)(=O)C)cc1
IUPAC Name N-[4-[1-hydroxy-2-(propan-2-ylamino)ethyl]phenyl]methanesulfonamide;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 308.82
Quantity 10 mg
Formula Weight 308.0961 g/mol