Få mere at vide
(±)-Sotalol-d7 HCl (iso-propyl-d7), CDN
Artikelnummer. 30325641
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30325641 0.005 g
30322312 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325641 Leverandør CDN Leverandørnr. CDND74110.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sotalol-D7 (isopropyl-D7) Hydrochloride
CAS 1398065-65-0
Molekylær formel C12 D7 H13 N2 O3 S . Cl H
Mængde 0.005 g
Synonym Sotalol D7 Hydrochloride (isopropyl-D7), Sotalol-D7 (isopropyl-D7) hydrochloride, N-[4-[2-(1,1,1,2,3,3,3-Heptadeuteriopropan-2-ylamino)-1-hydroxyethyl]phenyl]methanesulfonamide hydrochloride, Sotalol-d7 hydrochloride, N-[4-[1-Hydroxy-2-[[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]ethyl]phenyl]methanesulfonamide hydrochloride, 4'-[1-Hydroxy-2-(isopropyl-1,1,1,2,3,3,3-d7-amino)ethyl]methanesulfonanilide hydrochloride
InChI formel InChI=1 S/C12H20N2O3S.ClH/c1-9(2)13-8-12(15)10-4-6-11(7-5-10)14-18(3,16)17;/h4-7,9,12-15 H,8H2,1-3H3;1 H/i1D3,2D3,9 D;
SMIL Cl.[2 H]C([2 H])([2 H])C([2 H])(NCC(O)c1ccc(NS(=O)(=O)C)cc1)C([2 H])([2 H])[2 H]
IUPAC navn N-[4-[2-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)-1-hydroxyethyl]phenyl]methanesulfonamide;hydrochloride
Molekylvægt (g/mol) 315.868
Formel vægt 315.14 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity