Få mere at vide
Sorafenib-d3, TRC
Artikelnummer. 30426616
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30426616 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426616 Leverandør Toronto Research Chemicals Leverandørnr. TRCS67685210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sorafenib-methyl-d3
CAS 1130115-44-4
Renhedsgrad noter HPLC
Molekylær formel C21 2H3 H13 Cl F3 N4 O3
Mængde 10 mg
Synonym Sorafenib D3
InChI formel InChI=1S/C21H16ClF3N4O3/c1-26-19(30)18-11-15(8-9-27-18)32-14-5-2-12(3-6-14)28-20(31)29-13-4-7-17(22)16(10-13)21(23,24)25/h2-11H,1H3,(H,26,30)(H2,28,29,31)/i1D3
SMIL [2H]C([2H])([2H])NC(=O)c1cc(Oc2ccc(NC(=O)Nc3ccc(Cl)c(c3)C(F)(F)F)cc2)ccn1
IUPAC navn 4-[4-[[4-chloro-3-(trifluoromethyl)phenyl]carbamoylamino]phenoxy]-N-(trideuteriomethyl)pyridine-2-carboxamide
Molekylvægt (g/mol) 467.84
Formel vægt 467.1051
Procent renhed >95