Få mere at vide
Ansell™ Solvex™ 37-655 Series Green Nitrile Immersion Gloves
Shop alle Ansell produkter
Størrelse:
7
8
9
10
11
Pakningsstørrelse:
12 par
Beskrivelse
- Provides superb comfort for the wearer
- Food Approved
- No inner cotton flocking, reducing risk of lint contamination and making it ideal for production areas sensitive to the introduction of external impurities
- The sandpatch finish gives the glove a smooth surface that decreases indirect costs thanks to fewer rejects of fragile parts
Tekniske data
Tekniske data
| Materiale bagpå | Nitrile |
| Manchet materiale | Nitrile |
| Bedømmelse af skæremodstand | Level - 0 |
| Udvendig finish | Smooth |
| Produkttype | Immersion Gloves |
| Størrelse | 8 |
| Stil | 37-655 |
| Tykkelse (metrisk) | 0.38 mm |
| Handske manchet farve | Green |
| Handske manchet stil | Gauntlet |
| Vis mere |