missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Solvent Blue 38 (Technical Grade), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Solvent Blue 38 |
| CAS | 1328-51-4 |
| Molekylær formel | C24 H30 N4 O2 |
| Mængde | 5 g |
| Synonym | Acid Blue 87, Aizen Primula Turquoise Blue GL, Aizen Primula Turquoise Blue GLH, Amafast Turquoise 8GGL, Amanil Fast Turquoise, Amanil Fast Turquoise LB, Arlazol Fast Turquoise Blue 6GN, Atlantic Fast Turquoise LGA, Basacid Blue 752, Belamine Fast Turquoise LGL, C.I. 74180, C.I. Acid Blue 87, C.I. Solvent Blue 38, Calcodur Turquoise GL, Chlorantine Fast Turquoise Blue GLL, Chlorantine Fast Turquoise VLL, Chrome Leather Turquoise Blue GLL, Cibafix Direct Blue 86, Cuprofix Blue Green B, Cuprofix Blue Green FB, Cuprofix Turquoise Blue FBL, Daivougen Blue BF, Derma Fast Turquoise W 2GL, Diaphtamine Light Turquoise G, Diazol Light Turquoise JL, Direct Blue 86, Direct Emerald Blue GL, Direct Fast Blue L 7V, Direct Fast Turquoise, Direct Fast Turquoise Blue GL, Direct Fast Turquoise GL, Direct Light Turquoise Blue GL, Direct Light Turquoise GL, Direct Lightfast Blue Green GL, Direct Lightfast Emerald Blue GL, Direct Lightfast Turquoise, Direct Lightfast Turquoise Blue GL, Direct Lightfast Turquoise GL, Direct Lightfast Turquoise M, Direct Sun Green Blue GL, Direct Turquoise Blue GL, Disulfo copper phthalocyanine amine salt, Durazol Blue 8G, Durazol Blue 8GA, Durazol Blue 8GS, Durazol Fast Blue 8GS, Durazol Paper Blue 8G, Eliamina Light Turquoise G, Eliamina Light Turquoise GL, Fastogen Blue SBL, Fastusol Turquoise Blue LGA, Fenaluz Turquoise G, Finisol Blue Green G Heliogen Blue SBL, Heliogen Blue SBLA, Helion Turquoise Blue FGLL, Helion Turquoise Blue GLL, Hiltamine Turquoise Blue 8GL, Hispaluz Turquoise GE, Interchem Direct Fast Turquoise GLRC, Intrabond Liquid Turquoise LL, Intralite Turquoise GLL, Iragon Blue DBL 86, Kayarus Turquoise Blue GL, Kayaset Cyan J 301, Kiwa Turquoise Blue GL, Lampronol Fast Turquoise J, Light Fast Turquoise Blue G, Lightfast Turquoise, Lurantin Light Turquoise Blue GL, Lurantin Light Turquoise Blue GLD, Luxol Fast Blue MBS, Luxol Fast Blue MBSN, Methasol Fast Blue 2B, Methasol Fast Blue 2BX, Nankai Brilliant Fast Blue G, Neptune Blue 722, Nyanza Fast Turquoise GL, Nyastral Blue SS, Orbantin Turquoise Blue GL, Paliofast Blue SBL, Paper Fast Turquoise GP, Pergasol Turquoise Blue GAL Supra, Pergasol Turquoise G, Pheno Fast Turquoise GL, Pontamine Fast Turquoise 8GL, Saturn Turquoise Blue LG, Sepisol Fast Blue MBSN, Sirius Light Turquoise Blue GL, Sirius Supra Turquoise Blue GL, Sirius Supra Turquoise Blue Z-BGL, Sirius Turquoise GL, Solamine Light Turquoise Blue GL, Solantine Turquoise G, Solar Turquoise Blue GLL, Solius Light Turquoise Blue GL, Solophenyl Turquoise Blue GL, Solophenyl Turquoise Blue GL Supra, Solophenyl Turquoise Blue GTL, Solophenyl Turquoise Blue GTL Extra, Special Fast Turquoise 8GL, Spirit Soluble Fast Blue 6GHV, Sumilight Supra Turquoise Blue FB, Sumilight Supra Turquoise Blue G, Sumilight Supra Turquoise Blue G Conc., Symulon Direct Turquoise Blue B, Tertrodirect Fast Turquoise GL, Tetramine Fast D Extra Green, Toyo Cupro Cyanine Blue GL, Triantine Fast Turquoise GL, Triantine Light Turquoise Blue GL, Tricufix Turquoise Blue 2GL, Turquoise 8GL, Turquoise GLL, Vali Fast Blue 1605, [29H,31H-Phthalocyanine-C,C-disulfonato(4-)-κN29,κN30,κN31,κN32]-cuprate(2-) Disodium, [Dihydrogen phthalocyaninedisulfonato(2-)]-copper Disodium Salt, C.I. Direct Blue 86 |
| InChI formel | InChI=1S/C24H30N4O2/c29-23-5-1-3-21-19-9-17(13-27(21)23)11-25(15-19)7-8-26-12-18-10-20(16-26)22-4-2-6-24(30)28(22)14-18/h1-6,17-20H,7-16H2/t17-,18-,19+,20+/m0/s1 |
| SMIL | O=C1C=CC=C2[C@@H]3C[C@@H](CN(CCN4C[C@@H]5C[C@H](C4)C6=CC=CC(=O)N6C5)C3)CN12 |
| IUPAC navn | (1R,9S)-11-[2-[(1R,9S)-6-oxo-7,11-diazatricyclo[7.3.1.02,7]trideca-2,4-dien-11-yl]ethyl]-7,11-diazatricyclo[7.3.1.02,7]trideca-2,4-dien-6-one |
| Molekylvægt (g/mol) | 406.521 |
| Formel vægt | 406.237 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?