missing translation for 'onlineSavingsMsg'
Learn More

Solketal, 97%, Thermo Scientific Chemicals

Catalog Number p-193125
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
100-79-8
C6H12O3
132.16
MFCD00063238
RNVYQYLELCKWAN-UHFFFAOYSA-N
solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketal
7528
(2,2-dimethyl-1,3-dioxolan-4-yl)methanol
CC1(OCC(O1)CO)C
This item is not returnable. View return policy
100-79-8
C6H12O3
132.16
MFCD00063238
RNVYQYLELCKWAN-UHFFFAOYSA-N
solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketal
7528
(2,2-dimethyl-1,3-dioxolan-4-yl)methanol
CC1(OCC(O1)CO)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -50°C
Color Colorless
Density 1.0600g/mL
Boiling Point 188.0°C to 189.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4330 to 1.4350
Quantity 100 g
Beilstein 19, V,2, 362
Merck Index 15, 5259
Specific Gravity 1.06
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol, acetals, esters, ethers,, aromatic hydrocarbons, chlorinated hydrocarbons,, gasolines and petr.ether
Viscosity 11 mPa.s (20°C)
Formula Weight 132.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Solketal, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'