Få mere at vide
Solifenacin N-Oxide, TRC
Artikelnummer. 30011425
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011425 5 mg
30008985 50 mg
17968794 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011425 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6767105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 180272-28-0
Molekylær formel C23 H26 N2 O3
Molekylvægt (g/mol) 378.46
Synonym 2(1H)-Isoquinolinecarboxylic acid, 3,4-dihydro-1-phenyl-, (3R)-1-oxido-1-azabicyclo[2.2.2]oct-3-yl ester, (1S)-, Solifenacin N1-oxide
IUPAC navn [(3R)-1-oxido-1-azoniabicyclo[2.2.2]octan-3-yl] (1S)-1-phenyl-3,4-dihydro-1H-isoquinoline-2-carboxylate
SMIL [O-][N+]12CCC(CC1)[C@H](C2)OC(=O)N3CCc4ccccc4[C@@H]3c5ccccc5
Tekniske data
Kemisk navn eller materiale Solifenacin N-Oxide
Mængde 5 mg
InChI formel InChI=1S/C23H26N2O3/c26-23(28-21-16-25(27)14-11-18(21)12-15-25)24-13-10-17-6-4-5-9-20(17)22(24)19-7-2-1-3-8-19/h1-9,18,21-22H,10-16H2/t18?,21-,22-,25?/m0/s1
Formel vægt 378.1943