Få mere at vide
Solifenacin-d5 Hydrochloride, TRC
Artikelnummer. 30414710
Skift visning
Mængde:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30414710 1 mg
30423776 25 mg
30415958 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414710 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6767021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1426174-05-1
Molekylær formel C23 2H5 H21 N2 O2 . Cl H
Molekylvægt (g/mol) 403.96
Synonym (1S)-3,4-Dihydro-1-phenyl-d5-2(1H)-isoquinolinecarboxylic Acid (3R)-1-Azabicyclo-[2.2.2]oct-3-yl Ester Hydrochloride
IUPAC navn [(3R)-1-azabicyclo[2.2.2]octan-3-yl] (1S)-1-(2,3,4,5,6-pentadeuteriophenyl)-3,4-dihydro-1H-isoquinoline-2-carboxylate;hydrochloride
SMIL Cl.[2H]c1c([2H])c([2H])c([C@@H]2N(CCc3ccccc23)C(=O)O[C@H]4CN5CCC4CC5)c([2H])c1[2H]
Tekniske data
Kemisk navn eller materiale Solifenacin-d5 Hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C23H26N2O2.ClH/c26-23(27-21-16-24-13-10-18(21)11-14-24)25-15-12-17-6-4-5-9-20(17)22(25)19-7-2-1-3-8-19;/h1-9,18,21-22H,10-16H2;1H/t21-,22-;/m0./s1/i1D,2D,3D,7D,8D;
Formel vægt 403.2075