Få mere at vide
Solanidine, TRC
Artikelnummer. 30010283
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010283 1 mg
30008942 5 mg
30004614 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010283 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6824001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 80-78-4
Molekylær formel C27 H43 N O
Molekylvægt (g/mol) 397.64
Synonym Solanid-5-en-3-ol, (3β)-, Solanid-5-en-3β-ol (8CI), Solanidine (6CI,7CI), 1H-Naphth[2',1':4,5]indeno[1,2-b]indolizine, solanid-5-en-3-ol deriv., NSC 76025, Solanidin, Solatubin, Solatubine
SMIL C[C@H]1CC[C@@H]2[C@@H](C)[C@H]3[C@H](C[C@H]4[C@@H]5CC=C6C[C@@H](O)CC[C@]6(C)[C@H]5CC[C@]34C)N2C1
Tekniske data
Kemisk navn eller materiale Solanidine (Solatubine)
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C27H43NO/c1-16-5-8-23-17(2)25-24(28(23)15-16)14-22-20-7-6-18-13-19(29)9-11-26(18,3)21(20)10-12-27(22,25)4/h6,16-17,19-25,29H,5,7-15H2,1-4H3/t16-,17+,19-,20+,21-,22-,23+,24-,25-,26-,27-/m0/s1
Formel vægt 397.3345
Procent renhed >95%