Få mere at vide
Diversey™ Soft Care Wash H2 Extra Mild Cleanser
Volumen (metrisk):
500 ml
1 l
5 l
Emballage:
Dåse
Flaske med pumpe
Flaske med vippelåg
Flaske til dispenser
Beskrivelse
Soft Care Wash is an ultra-mild, hypoallergenic hand, hair and body washing liquid. Soft Care Wash has been specifically developed for sensitive skin. It is free of irritants, allergens and other unwanted substances. This eco-certified product is suitable for use in all areas, but especially in healthcare, leisure and food service.
Key properties
- Excellent for cleaning hands, body and hair
- Surfactant system effectively removes dirt and grease
- Ultra mild profile with moisturizers to promote safe & healthy skin
- Fulfills the criteria of the Nordic Ecolabel scheme
- Skin compatibility tested (Patch test)
- Hypoallergenic; endorsed by Nordic Asthma & Allergy Foundation
- Dye-free
- Does not contain fragrance, thus eliminating the risk of tainting food
- Free from undesirable ingredients such as Cocamide DEA, parabens and phosphates
- Easy to rinse
Benefits
- Effective cleaning from top to toe: hands, body, and hair
- Healthy skin even with frequent washing
- Safe for use in food service, food preparation, and healthcare environments
- Proven skin compatibility and best possible safety profile for increased user acceptance
Tekniske data
Tekniske data
| Produkttype | Extra Mild Cleanser |
| Emballage | Flaske med pumpe |
| Mængde | 10 Pk. |
| Volumen (metrisk) | 500 ml |