missing translation for 'onlineSavingsMsg'
Learn More
Sofosbuvir, MedChemExpress Product Code.: 16492407

Sofosbuvir, MedChemExpress

Product Code. p-200096097
50 mg, 50mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16492407

Brand: Medchem Express HY15005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sofosbuvir (GS-7977) is an HCV RNA replication inhibitor with an EC50 of 92 nM.

Sofosbuvir (GS-7977) is an HCV RNA replication inhibitor with an EC50 of 92 nM.

Specifications

Chemical Name or Material Sofosbuvir
CAS 1190307-88-0
Color Brown
Purity Grade Notes Research
Molecular Formula C22H29FN3O9P
Quantity 50 mg
Synonym GS-7977 PSI-7977
Solubility Information DMSO : 100 mg/mL (188.88 mM; Need ultrasonic) ∣H2O : 25 mg/mL (47.22 mM; ultrasonic and warming and heat to 50°C)
SMILES O=C1N([C@H]2[C@]([C@H](O)[C@@H](CO[P@](OC3=CC=CC=C3)(N[C@@H](C)C(OC(C)C)=O)=O)O2)(C)F)C=CC(N1)=O
Molecular Weight (g/mol) 529.45
Formula Weight 529.45
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Sofosbuvir, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.