Få mere at vide
Notrax Sof-Tred™ Microcellular Vinyl Foam with Pebble Pattern, Blue
Artikelnummer. 17712088
Artikelnummer. 17712088 Leverandør Notrax Leverandørnr. 411S0323BU
Notrax Sof-Tred™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit.
  • Pebble pattern provides non-directional traction and is easy to clean
  • Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
  • All four sides are bevelled and sealed to minimize tripping hazards
Tekniske data
Farve Blue
Længde (engelsk) 36 in.
Længde (metrisk) 91 cm
Materiale Microcellular Vinyl
Type Industrial and Commercial Matting
Bredde (metrisk) 60 cm