Learn More
Sodium Tungstate Dihydrate, Supelco™
Product Code. 15864900
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15864900 250 g 250g
15807520 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15864900 Supplier Supelco Supplier No. 1066730250
 Restricted Item
Forventet forsendelse: 19-11-2026
For analysis EMSURE™
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10213-10-2
Specifications
Melting Point 100°C (Elimination of water of crystallisation)
Quantity 250 g
MDL Number MFCD00149190
Synonym Sodium tungstate dihydrate, Sodium tungstate dihydrate
Molecular Weight (g/mol) 329.85
Grade Research
Density 3.23 g/cm3 at 20°C
Chemical Name or Material Sodium Tungstate Dihydrate