missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulphate Solution, 0.1M, Fisher Chemical™
Click to view available options
Quantity:
100 mL
1000 mL
Specifications
Concentration | 0.1M |
Quantity | 100 mL |
Chemical Name or Material | Sodium Thiosulphate Solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'