missing translation for 'onlineSavingsMsg'
Få mere at vide

Sodium Thiosulfate Pentahydrate, TRC

Artikelnummer. 30412962
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30412962 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30412962 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6736451G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Sodium Thiosulfate hydrate
CAS 128114-51-2
Molekylær formel 2Na . O3 S2 . x(H2 O)
Mængde 1 g
Synonym Thiosulfuric Acid (H2S2O3) Disodium Salt Hydrate, Thiosulfuric Acid Disodium Salt Hydrate, Disodium Thiosulfate Hydrate, Disodium Thiosulphate Hydrate, Hypo Sodium Thiosulfate (Na2S2O3) Hydrate
InChI formel InChI=1S/2Na.H2O3S2.H2O/c;;1-5(2,3)4;/h;;(H2,1,2,3,4);1H2/q2*+1;;/p-2
SMIL O.[Na+].[Na+].[O-]S(=O)(=S)[O-]
IUPAC navn disodium;dioxido-oxo-thioxo-$l^{6}-sulfane;hydrate
Molekylvægt (g/mol) 176.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.