missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, Honeywell
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Description
- ≥95% for general laboratory use.
Specifications
Quantity | 2.5 kg |
Linear Formula | Na2S2O3 |
UN Number | NONH for all modes of transport |
IUPAC Name | disodium pentahydrate sulfanidesulfonate |
Formula Weight | 158.11g/mol |
Percent Purity | 99% |
Grade | ReagentPlus™ |
Chemical Name or Material | Sodium Thiosulfate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'