Få mere at vide
Sodium Thiocyanate solution 1mol/L, Fisher Chemical™
Artikelnummer. 12993624 Shop alle Fisher Chemical produkter
Skift visning
Mængde:
1000 mL
Pakningsstørrelse:
1000ml
Artikelnummer. Mængde Pakningsstørrelse
12993624 1000 mL 1000ml
1 options
Artikelnummer. 12993624 Leverandør Fisher Chemical Leverandørnr. VL 312 1000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Tekniske data
Kemisk navn eller materiale Sodium Thiosulphate
Emballage Bottle
CAS 57-55-6
Molekylær formel C3H8O2
Koncentration 1M (1N)
InChI nøgle DNIAPMSPPWPWGF-UHFFFAOYSA-N
SMIL CC(CO)O
IUPAC navn propane-1,2-diol
Molekylvægt (g/mol) 76.095
PubChem CID 1030
Mængde 1000 mL
ChEBI CHEBI:16997