missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiocyanate, 98 to 102% (titration), Reagent Grade, Honeywell™
Click to view available options
Quantity:
250 g
1 kg
3 kg
Packaging:
Poly bottle
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | 308°C |
Color | Colorless |
Physical Form | Solid |
Quantity | 1 kg |
Linear Formula | NaSCN |
UN Number | NONH for all modes of transport |
Beilstein | 3594965 |
IUPAC Name | sodium cyanosulfanide |
Formula Weight | 81.07g/mol |
Percent Purity | 98 to 102% (titration) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'