missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Natriumtetraborat, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Disodium tetraborate |
| CAS | 1330-43-4 |
| Mængde | 5 g |
| Molekylær formel | B4 O7 . 2Na |
| Synonym | Sodium tetraborate, Boron sodium oxide (B4Na2O7) (9CI','ACI), Boric acid (H2B4O7)','disodium salt (8CI), Sodium tetraborate (Na2B4O7) (7CI), Anhydrous borax, Borax glass, DC Dehybor, Dehybor, Ecoboron PRO, Etibor 68, Fireless B, Fireless B Liquid, FR 28, FR 28 (borate), Fused Borax, Granubor, Rasorite 65, Sodium biborate, Sodium borate, Sodium boron oxide (Na2B4O7), Spraybor |
| InChI formel | InChI=1S/B4O7.2Na/c5-1-7-3-9-2(6)10-4(8-1)11-3;;/q-2;2*+1 |
| SMIL | [Na+].[Na+].[O-]B1OB2OB([O-])OB(O1)O2 |
| IUPAC navn | disodium; 3,7-dioxido-2,4,6,8,9-pentaoxa-1,3,5,7-tetraborabicyclo[3.3.1]nonan |
| Molekylvægt (g/mol) | 201.22 |
| Formel vægt | 201.9812 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?