Få mere at vide
Sodium Starch Glycolate, TRC
Product Code. 30132743
Change view
Click to view available options
Mængde:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30132743 100 mg
30133810 1 g
17997714 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132743 Supplier Toronto Research Chemicals Supplier No. TRCS666990100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9063-38-1
Synonym Carboxymethyl Starch Sodium Salt, Covagel, Crayon B 300P, Deprogel, Emsize CMS 100, Emsize CMS 60, Estarl A 100, Explosol, Explotab, Explotab CL-V, Expotab PCF, F 500 Papeal No. 50, Glycol YS, Glycolys LV, Glycolys Std, KMK-BUR 1, KMK-OK, Kiprogum F 500, LCW Covagel, Libeng, P 5000, Papeal F 500 No. 50, Polvitex Z, Polytex 60, Primojel, Sodium CM-starch, Sodium Carboxymethyl Starch, Solvitose CL, Vivastar P, Vivastar P 3500, Vivastar P 5000, WS 1R, Walocel CRT 100GA, USP Sodium Starch Glycolate Type A
Specifications
Kemisk navn eller materiale Sodium Starch Glycolate Type A