Learn More
Sodium Propyl Parahydroxybenzoate, Mikromol™
Product Code. 17973636
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
17973636 100 mg 100mg
1 options
Product Code. 17973636 Supplier Mikromol Supplier No. MM0432.01
 Restricted Item
Forventet forsendelse: 27-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Sodium Propyl Parahydroxybenzoate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sodium Propyl Parahydroxybenzoate
CAS 35285-69-9
Analyte or Component Names Sodium Propyl Parahydroxybenzoate
Shipping Condition Room Temperature
Molecular Formula C10 H11 O3 . Na
Certifications/Compliance ISO 17034
Synonym Sodium Propyl Parahydroxybenzoate
InChI Formula InChI=1S/C10H12O3.Na/c1-2-7-13-10(12)8-3-5-9(11)6-4-8;/h3-6,11H,2,7H2,1H3;/q;+1/p-1
SMILES [Na+].CCCOC(=O)c1ccc([O-])cc1
IUPAC Name sodium;4-propoxycarbonylphenolate
Molecular Weight (g/mol) 202.18
Quantity 100 mg
Formula Weight 202.0606 g/mol