Få mere at vide
Sodium Propionate-3,3,3-d3, CDN
Artikelnummer. 30324997
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324997 1 g -
30321102 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324997 Leverandør CDN Leverandørnr. CDND77481G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sodium Propionate-3,3,3-D3
CAS 1210410-35-7
Molekylær formel C3 D3 H2 O2 . Na
Mængde 1 g
Synonym Propanoic-3,3,3-d3 acid, sodium salt (1:1) (ACI), Sodium propionate-3,3,3-D3, Sodium propanoate-3,3,3-d3, Propanoic-3,3,3-d3 acid sodium salt
InChI formel InChI=1 S/C3H6O2.Na/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1/i1D3;
SMIL [Na+].[2 H]C([2 H])([2 H])CC(=O)[O-]
IUPAC navn sodium;3,3,3-trideuteriopropanoate
Molekylvægt (g/mol) 99.079
Formel vægt 99.038 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity