Få mere at vide
Sodium Pregnenolone-17alpha,21,21,21-d4 Sulfate, TRC
Artikelnummer. 30414110
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30414110 5 mg
30411467 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414110 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7122215MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1485492-21-4
Molekylær formel C21H27D4NaO5S
Molekylvægt (g/mol) 422.55
Synonym (3β)-3-(Sulfooxy)pregn-5-en-20-one-17α,21,21,21-d4 sodium salt, 3β-Hydroxypregn-5-en-20-one-17α,21,21,21-d4 hydrogen sulfate sodium salt, Pregnenolone-17α,21,21,21-D4 sulfate sodium salt, Pregnenolone-3-sulfate-17α,21,21,21-d4 sodium salt, Pregnenolone-17α,21,21,21-d4 3β-sulfate sodium salt, Pregnenolone-17α,21,21,21-d4 sodium sulfate, 3β-Hydroxypregn-5-en-20-one-17α,21,21,21-d4 sulfate sodium salt, Sodium Pregnenolone-17α,21,21,21-d4 sulfate, Sodium Pregnenolone-17alpha,21,21,21-d4 Sulfate
IUPAC navn sodium (3S,8S,9S,10R,13S,14S,17S)-17-(acetyl-d3)-10,13-dimethyl-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl-17-d sulfate
SMIL C[C@@](C1=CC2)(CC[C@H](O[S](=O)(O[Na])=O)C1)[C@]3([H])[C@]2([H])[C@@](CC[C@]4([2H])C(C([2H])([2H])[2H])=O)([H])[C@]4(C)CC3
Tekniske data
Kemisk navn eller materiale Pregnenolone-17alpha,21,21,21-D4 Sulfate Sodium Salt
Mængde 5 mg
InChI formel InChI=1S/C21H32O5S.Na/c1-13(22)17-6-7-18-16-5-4-14-12-15(26-27(23,24)25)8-10-20(14,2)19(16)9-11-21(17,18)3;/h4,15-19H,5-12H2,1-3H3,(H,23,24,25);/q;+1/p-1/t15-,16-,17+,18-,19-,20-,21+;/m0./s1/i1D3,17D;
Formel vægt 422.2