Learn More
Sodium phosphate monobasic monohydrate, ACS Reagent, Honeywell Fluka™
Catalog Number p-7072321
Click to view available options
Quantity:
25 g
250 g
500 g
1 kg
3 kg
6 x 1 kg
10049-21-5
H5NaO5P
139.00
MFCD00149208
BBMHARZCALWXSL-UHFFFAOYSA-N
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
516949
O.[Na+].OP(O)(O)=O
10049-21-5
H5NaO5P
139.00
MFCD00149208
BBMHARZCALWXSL-UHFFFAOYSA-N
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
516949
O.[Na+].OP(O)(O)=O
ACS Reagent,≥98%
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 25 g
Linear Formula NaH2PO4 · H2O
UN Number NONH for all modes of transport
IUPAC Name sodium phosphoric acid hydrate
Formula Weight 137.99g/mol
Percent Purity 98 to 102%, ≥98%
Grade ACS Reagent
Chemical Name or Material Sodium phosphate monobasic monohydrate
Hazard Statement
  • EUH210-Safety data sheet available on request.
Precautionary Statement