Learn More
Sodium phosphate monobasic monohydrate, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
25 g
250 g
500 g
1 kg
3 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 25 g |
Linear Formula | NaH2PO4 · H2O |
UN Number | NONH for all modes of transport |
IUPAC Name | sodium phosphoric acid hydrate |
Formula Weight | 137.99g/mol |
Percent Purity | 98 to 102%, ≥98% |
Grade | ACS Reagent |
Chemical Name or Material | Sodium phosphate monobasic monohydrate |
Safety and Handling
Hazard Statement
- EUH210-Safety data sheet available on request.