Få mere at vide
Sodium Pentanoate-d9, CDN
Artikelnummer. 30327060
Skift visning
Mængde:
0.25 g
Artikelnummer. Mængde
30327060 0.25 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327060 Leverandør CDN Leverandørnr. CDND82360.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sodium Pentanoate-D9
CAS 2964559-19-9
Molekylær formel C5 D9 O2 . Na
Mængde 0.25 g
Synonym Sodium Pentanoate-D9, Pentanoic-d9 acid sodium salt, Pentanoic acid-d9 sodium salt, Valeric acid-d9 sodium salt, Valeric-d9 acid sodium salt, Sodium valerate-d9
InChI formel InChI=1 S/C5H10O2.Na/c1-2-3-4-5(6)7;/h2-4H2,1H3,(H,6,7);/q;+1/p-1/i1D3,2D2,3D2,4D2;
SMIL [Na+].[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)[O-]
IUPAC navn sodium;2,2,3,3,4,4,5,5,5-nonadeuteriopentanoate
Molekylvægt (g/mol) 133.169
Formel vægt 133.1065 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity