Learn More
Sodium Oxalate, ≥99.5%, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | NaOOCCOONa |
UN Number | NONH for all modes of transport |
Beilstein | 3631622 |
Formula Weight | 134g/mol |
Percent Purity | ≥99.5% |
Grade | ACS Reagent |
Packaging | Poly Bottle |
Chemical Name or Material | Sodium Oxalate |