Få mere at vide
Sodium O,O-Dimethyl Thiophosphate (Technical grade), TRC
Artikelnummer. 30136311
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136311 1 g
30134968 500 mg
17933855 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136311 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6242401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 23754-87-2
Molekylær formel C2 H6 O3 P S . Na
Molekylvægt (g/mol) 164.1
Synonym Phosphorothioic acid, O,O-dimethyl ester, sodium salt (1:1), Methyl sodium phosphorothioate ((MeO)2(NaO)PS) (6CI), Phosphorothioic acid, O,O-dimethyl ester, sodium salt (8CI,9CI), Sodium O,O-dimethyl phosphorothioate, Sodium O,O-dimethyl thiophosphate, O,O-Dimethylphosphorothioic acid sodium
IUPAC navn sodium;[methoxy(sulfido)phosphoryl]oxymethane
SMIL [Na+].COP(=O)([S-])OC
Tekniske data
Kemisk navn eller materiale O,O-Dimethylphosphorothioic acid sodium salt
Mængde 1 g
InChI formel InChI=1S/C2H7O3PS.Na/c1-4-6(3,7)5-2;/h1-2H3,(H,3,7);/q;+1/p-1
Formel vægt 163.9673