missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitroprusside Dihydrate, (Crystalline),MP Biomedicals
Click to view available options
Quantity:
100 g
Specifications
Physical Form | Crystalline |
Quantity | 100 g |
IUPAC Name | disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate |
Formula Weight | 297.95 |
Chemical Name or Material | Sodium Nitroprusside Dihydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'