Learn More
Sodium nitrite, 98.5%, for analysis Product Code.: 10339333
Sodium nitrite, 98.5%, for analysis
Product Code. 10339333 Shop All Thermo Scientific Chemicals Products
5 kg, Plastic bottle, 5kg
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
5kg
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 10339333
Brand: Thermo Scientific Acros 217600050
Request Bulk or Custom Format
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
CAS: 7632-00-0 | NNaO2 | 69.00 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium;nitrite
SMILES [Na+].[O-]N=O
Specifications
Color Yellow
Physical Form Crystalline Powder and/or Crystals
Quantity 5 kg
Assay Percent Range 98.5%
Linear Formula NaNO2
Fieser 01,1097; 02,386; 04,459; 06,547; 09,432; 11,491; 13,170; 16,86
Merck Index 15, 8779
Formula Weight 69
Percent Purity ≥98.0%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Sodium nitrite
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0470544 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code217600050,217600010
Lot NumberA0472286 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code217600050,217600010
Lot NumberA0467256 Certificate TypeCertificate of Analysis Date31/01/2025 Product Code217600050,217600010
Lot NumberA0458489 Certificate TypeCertificate of Analysis Date22/12/2023 Product Code217600050,217600010
Lot NumberA0448366 Certificate TypeCertificate of Analysis Date14/12/2023 Product Code217600050,217600010
RUO – Research Use Only
Product Content Correction
Product Title
Sodium nitrite, 98.5%, for analysis >