Learn More
Sodium Nitrite 0.5 M (0.5 N) Analytical Volumetric Solution (AVL), Reagecon
Shop All Reagecon Products
Click to view available options
Quantity:
1 L
5 L
Packaging:
HDPE Bottle GL45
HDPE Container
Unit Size:
1 L
5 L
Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- Accurate, stable, reproducible and fully factorized.
- Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
- Stability is proven (either opened or unopened) for the shelf life of this product.
- Certified to a specification of ± 0.1%.
- Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
- May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
Specifications
Specifications
Description | Accuracy ± 0.1% |
Packaging | HDPE Bottle GL45 |
Physical Form | Liquid |
Concentration | 0.5 M |
Technique | Titration |
Quantity | 1 L |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
NANO0525E1 | Certificate of Analysis | 31/05/2025 | NANO051 |
NANO0524M1 | Certificate of Analysis | 29/01/2025 | NANO051 |
NANO0523M1 | Certificate of Analysis | 07/02/2024 | NANO051 |
NANO0523B1 | Certificate of Analysis | 03/05/2023 | NANO051 |
NANO0521M1 | Certificate of Analysis | 09/03/2022 | NANO051 |
Product Content Correction
Product Title
Sodium Nitrite 0.5 M (0.5 N) Analytical Volumetric Solution (AVL), Reagecon >
Spot an opportunity for improvement?Share a Content Correction