Learn More
Sodium Nitrate, Supelco™
Product Code. 15874850
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15874850 1 kg 1kg
15864850 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15874850 Supplier Supelco Supplier No. 1065371000
 Restricted Item
Forventet forsendelse: 26-11-2026
For analysis EMSURE™ ACS,ISO,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7631-99-4
Specifications
Melting Point 308°C
Quantity 1 kg
MDL Number MFCD00011119
Synonym Sodium nitrate, Nitric acid sodium salt, Sodium nitrate
Molecular Weight (g/mol) 84.99
Grade Research
Density 2.26 g/cm3 at 20°C
Chemical Name or Material Sodium Nitrate