Learn More
Sodium nitrate, Puratronic™, 99.999% (metals basis)
Product Code. 11337037
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337037 25 g 25g
11317037 5 g 5g
11327037 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11337037 Supplier Thermo Scientific Alfa Aesar Supplier No. 010873.14
 Restricted Item
På lager
Add to basket
CAS: 7631-99-4 ∣ NNaO3 ∣ 84.99 g/mol
Sodium nitrate is used in the production of fertilizers, nitric acid, pyrotechnics, smoke-bombs, glass and pottery enamels. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent. Further it finds use as food preservative and as a solid rocket propellant. It is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
PubChem CID 24268
ChEBI CHEBI:63005
IUPAC Name sodium nitrate
SMILES [Na+].[O-][N+]([O-])=O
Specifications
Melting Point 308°C
Physical Form Powder
Quantity 25 g
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystals
Assay Percent Range 99.999% (metals basis)
Density 2.26 g/cm3 at 20°C
Solubility Information Freely soluble in water,ammonia and hydrazine; soluble in alcohol; slightly soluble in pyridine.
Formula Weight 84.99
Percent Purity 100%
Grade Ungraded
Chemical Name or Material Sodium nitrate