Learn More
Sodium methyl sulfate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 512-42-5 |
| Molecular Formula | CH3NaO4S |
| MDL Number | MFCD00013457 |
| Quantity | 25 g |
| SMILES | O=S(OC)([O-])=O.[Na+] |
| Molecular Weight (g/mol) | 134.09 |
| Percent Purity | 98% |
| Chemical Name or Material | Sodium methyl sulfate |